Learn More
Honeywell Uvex™ Genesis™ Reading Magnifier Safety Glasses
High-impact protection
Manufacturer:  Honeywell S3771
 View more versions of this product
Catalog No. 18-813-401
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ANSI Z87.1-2010 and CSA Z94.3 standards
Black
+1.50
99.90%
Scratch-resistant
Minimizes outdoor sunlight and glare
Ultra-dura Anti-scratch
Reading Magnifier Spectacles
Gray