Honeywell Uvex™ Genesis™ Reading Magnifier Safety Glasses
High-impact protection
Manufacturer: Honeywell S3775
 View more versions of this product
Catalog No. 18-813-405
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Certifications/Compliance ANSI Z87.1-2010 and CSA Z94.3 standards
    For Use With (Application) For varying light conditions
    Frame Color Black
    Lens Coating ultra dura anti scratch
    Lens Tint SCT-Reflect 50
    Magnification Power 1
    Surface Coating Ultradura antiscratch
    UV Protection 99.90%
    Type Reading Magnifier Spectacles
  • Description & Specifications
Related Products
Product Content Feedback