Honeywell Uvex™ Genesis™ Reading Magnifier Safety Glasses
High-impact protection
Manufacturer: Honeywell S3776
 View more versions of this product
Catalog No. 18-813-406
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ANSI Z87.1-2010 and CSA Z94.3 standards
    For Use With (Application) For varying light conditions
    Frame Color Black
    Lens Coating ultra dura anti scratch
    Lens Tint SCT-Reflect 50
    Magnification Power 1.5
    Surface Coating Ultradura antiscratch
    UV Protection 99.90%
    Type Reading Magnifier Spectacles
  • Description & Specifications
Related Products
Product Content Feedback