Uvex™ Genesis™ Safety Glasses, Replacement Lenses
Manufacturer: Honeywell Safety Products S6906
 View more versions of this product
Catalog No. 19-148-320
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) genesis Spectacles
    Lens Tint Shade 2
    Surface Coating Ultradura hardcoat
    Type Replacement Lenses/Spectacles
  • Description & Specifications
Related Products
Product Content Feedback