Learn More
Uvex™ Genesis X2™ Safety Glasses
Sport styled wraparound lens provides exceptional peripheral protection
Manufacturer:  Honeywell Safety Products S3500
 View more versions of this product
Catalog No. 17-377-419
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lifetime frame guarantee
Scratch-resistant
Exceed ANSI Z87.1-2003 standards; meet stringent military V0 ballistic impact requirements.
Polycarbonate
Ratcheting Spatula
Clear
Provides eye protection in a variety of industries
Polycarbonate with elastomer
Silver; Navy
Wraparound