Learn More
Honeywell Safety Products™ Uvex™ Hypershock™ Safety Eyewear
Delivers ultimate combination of high-performance protection and aggressive sport inspired styling
Manufacturer:  Honeywell Safety Products™ S2960XP
 View more versions of this product
Catalog No. 19-066-215
Add to cart
Description
Description
    • Streamlined padded temples and molded nosepiece
    • Designed with anti-fog lens coating for long-lasting fog free vision
    • Provides 2X scratch-resistance
    • 100% dielectric
    • Integrated hinge mechanisms with dual cam action
    • Soft, molded inserts along temple provide comfortable and secure fit
    • Lightweight, wrap-around frame provides increased coverage, side protection and superior peripheral vision
Compliance
Meet ANSI Z87.1-2010 standards.
Specifications
Specifications
Safety
Smoke Brown
Anti-fog, Anti-static, Anti-UV, Scratch-resistant
Agriculture, Construction, Food Services, Manufacturing, Oil and Gas, Steel and Metals, Transportation, Wind Energy industries
Clear
Safety Eyewear