Learn More
Honeywell Safety Products™ Uvex™ Hypershock™ Safety Eyewear
Delivers ultimate combination of high-performance protection and aggressive sport inspired styling
Manufacturer:  Honeywell Safety Products™ S2970XP
 View more versions of this product
Catalog No. 19-066-220
Add to cart
Description
Description
  • Streamlined padded temples and molded nosepiece
  • Designed with anti-fog lens coating for long-lasting fog free vision
  • Provides 2X scratch-resistance
  • 100% dielectric
  • Integrated hinge mechanisms with dual cam action
  • Soft, molded inserts along temple provide comfortable and secure fit
  • Lightweight, wrap-around frame provides increased coverage, side protection and superior peripheral vision
Applications:
Indoor and outdoor applications in construction, oil, gas and chemical, utilities, landscaping and manufacturing.
Compliance
Meet ANSI Z87.1-2010 standards.
Specifications
Specifications
Safety
Clear Ice
Anti-fog, Anti-static, Anti-UV, Scratch-resistant
Agriculture, Construction, Food Services, Manufacturing, Oil and Gas, Steel and Metals, Transportation, Wind Energy industries
Clear