Learn More
Honeywell Safety Uvex? Livewire? Sealed Eyewear
For protecting workers against intense wind, sun, dirt, dust, debris, as well as high heat and humidity
Manufacturer:  Honeywell Safety S2607XP
 View more versions of this product
Catalog No. 19-039-926
  Check Availability
Add to cart
Alert:
Not for chemical splash applications
Specifications Description & Specifications
Specifications
Sealed eyewear
Anti-fog,Anti-static,Anti-UV,IR Shade 3,Scratch-resistant
Agriculture, Construction, Forestry, Manufacturing, Mining, Municipal Services, Oil and Gas, Steel and Metals, Utilities, Welding, Wind Energy
Black, matte