Uvex™ Millennia™ Protective Safety Glasses
Futuristic wraparound styling with exceptional coverage and protection
Manufacturer: Honeywell Safety Products 11150351
 View more versions of this product
Catalog No. 19-039-209
  Check Availability
Add to Cart
    Specifications
    Frame Color Black
    Lens Coating UD
    Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3 standards as well as military V0 ballistic test for impact.
    Lens Material High-impact, Abrasion-resistant Polycarbonate
    Lens Tint TSR Gray
  • Description & Specifications
Related Products
Product Content Feedback