Learn More
Uvex™ Millennia™ Protective Safety Glasses
Futuristic wraparound styling with exceptional coverage and protection
Manufacturer:  Honeywell Safety Products 11150353
 View more versions of this product
Catalog No. 19-039-211
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
SCT-Reflect 50
Meet ANSI Z87.1-2003 and CSA Z94.3 standards as well as military V0 ballistic test for impact.
Black
Impact-resistant,Scratch-resistant
Polycarbonate