Uvex™ PrecisionPro™ Eyewear
Ideal for quality control, inspection, technical applications, lab work, aerospace, fine maintenance and mechanics
Manufacturer: Honeywell Safety Products SX0202
 View more versions of this product
Catalog No. 18-813-458
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Lens Coating Supra-Dura Anti-scratch
    For Use With (Application) For low-light applications where enhanced contrast is needed
    Certifications/Compliance Exceeds ANSI Z87.1-2003.
    Lens Tint Amber
  • Description & Specifications
Related Products
Product Content Feedback