Learn More
Honeywell™ Uvex™ Protege™ Safety Glasses
Ultra-lightweight and soft temple tip pads for a secure, comfortable fit
Supplier: Honeywell™ S4212
This item is not returnable.
View return policy
Description
- Secure, wraparound brow accommodates a wide range of users
- Proprietary Floating Lens™ design offers increased flexibility for better fit
- Soft, clear nose pads prevent slipping
uvextra™ Anti-Fog Coating (AF)
- Permanently bonded to lens
- Minimizes fogging
- Permanently bonded to lens
- Scratch- and chemical-resistant
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Warranty and Services
Lifetime frame guarantee.
Specifications
ANSI Z87.1-2015, CSA Z94.3-2015, Military V0 Ballistic Test | |
Scratch-resistant, Impact-resistant | |
Sandstone | |
Polycarbonate |
Lifetime frame guarantee | |
SCT-Reflect 50 | |
Cushioned |