Learn More
Honeywell™ Uvex™ Replacement Lenses for Flex Seal™ Goggles
Lightweight, silicone body conforms to assure the highest level of comfort and eye protection. Exceptional chemical splash and impact resistance. Fits over most prescription eyewear.
Supplier: Honeywell™ S710X
Description
Uvex Flex Seal goggle is made of a lightweight, silicone body, which conforms to the face, to assure the highest level of comfort and eye protection. The silicone body stays supple even in extreme temperatures and seals to almost any face shape, while offering exceptional all-day comfort.
- Available in HydroShield or Uvextreme Anti-Fog lens coatings
- Fits comfortably over prescription eyewear
- Available in various lens tints
- Economical lens replacement
Specifications
Clear | |
Replacement lens | |
Flex Seal Goggles |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.