Learn More
Honeywell™ Uvex™ Skyline™ Safety Glasses
Slimmer style for smaller faces
Manufacturer:  Honeywell™ S2000
Catalog No. 19-050-900
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Wraparound unilens design is 10mm slimmer than skyper™ spectacles—ideal for normal to slender profiles
  • Offers the same protection and comfort as the skyper™, including molded-in nosebridge, sideshields, and browguard
  • Adjustable lens inclination and temples provide maximum comfort
  • Easy-to-replace, impact-resistant polycarbonate lens blocks out 99.9% of harmful UVA and UVB radiation
Order Info
Available with choice of Ultradura™ hardcoat (UD) or Uvextreme™ AF (XTR) antifog, antiscratch, and antistatic lens coatings
Compliance
Meet ANSI Z87.1-2003 and CSA Z94.3-1999 standards.
Specifications
Specifications
Safety
Documents
Accessories