Uvex™ Skyper™ X2 Safety Glasses
Simplicity of a unilens with sleek, sporty appeal
Manufacturer: Honeywell Safety Products S2810X
 View more versions of this product
Catalog No. 19-039-007
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 
    Specifications
    Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3-1999 standards.
    Lens Material Impact-resistant Polycarbonate
    Lens Tint Clear
    Frame Color Black
    Lens Coating Uvextreme Anti-fog
    Temple Style Duoflex
    • Impact-resistant wraparound polycarbonate lenses block 99.9% of harmful UVA and UVB radiation
    • Black frames available with Ultradura™ hardcoat (UD) or Uvextreme™ AF (XTR) antifog, antiscratch, and antistatic lens coatings
    • Molded-in nosebridge for comfort and safety
    • Molded-in browguard reduces the danger of flying particulates
    • Patented, adjustable, comfort-cushioned Duoflex™ temples for custom fit with superior peripheral protection for a variety of work applications
    • Ratcheting lens inclination
    Replacement lenses available on our web site or the Fisher HealthCare catalog.
  • Description & Specifications
Related Products
Product Content Feedback