Learn More
Honeywell Safety Products™ Uvex™ Stealth™ Chemical-Splash Goggles
Sleek, low-profile design fits well with respirators, hardhats or welding helmets
Manufacturer:  Honeywell Safety Products™ S3960CI
 View more versions of this product
Catalog No. 19-823-231
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
Sleek, low-profile design fits well with respirators, hardhats or welding helmets
  • Polycarbonate lens is coated with uvextreme™ anti-fog coating (XTR) for protection against fogging, scratches, static dust, and 99.9% of harmful UV rays up to 385nm
  • Toric lens provides superior optics and improves peripheral vision
  • Combines durable polypropylene frame and soft, flexible elastomer inner flange
  • Provides superior splash protection
  • Indirect ventilation system directs airflow over the lens to minimize fogging and improve vision—without compromising splash and impact protection
  • Neoprene headband is textured to prevent slipping and features quick-adjust clips for easy and secure fit
  • Snap-off/snap-on lens replacement system suits many applications and environments
Order Info
Visit www.fishersci.com (keyword: uvex stealth) for replacement lenses and Rx carrier adapter that allows prescription glasses to be used with goggles.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
Specifications
Specifications
Anti-fog
Gray
Uvextreme Anti-fog
Clear
Neoprene headband