Uvex™ Stealth™ Chemical-Splash Goggles
Sleek, low-profile design fits well with respirators, hardhats or welding helmets
Manufacturer: Honeywell Safety Products S39611C
 View more versions of this product
Catalog No. 18-999-2316
  Check Availability
Add to Cart
    Specifications
    Lens Tint Gray
    Material Neoprene headband
    Surface Coating Uvextreme Antifog
    Ventilation Details Indirect ventilation system
    Lens Coating Uvextreme Antifog
    Type Chemical Splash Goggles
    Certifications/Compliance ANSI Z87.1-2003 and CSA Z94.3 standards
    Color Teal/Light Gray Body
    Frame Color Teal/Light Gray
    Frame Material Polypropylene
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback