Learn More
Uvex™ Stealth™ Goggle Products for Science Education
Chemical splash goggles offer comfort and protection with a close fit for superior optics
$22.50
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
S47614 Products for Science Education
View Documents
Honeywell Safety Products
S3960C/EMD
N/A Gray Body, Neoprene Band
Each for $22.50
Case of 50 EA for $1,125.00
Add to cart
Description
Description
  • Uvextreme™ AF anti-fog lens coating
  • Durable polypropylene frame
  • Soft flexible elastomer seals comfortably to facial contours
  • Unique system makes lens replacement a snap
  • Improved indirect ventilation system also reduces fogging
Compliance
Meets ANSI Z87.1-2010 standards.