Uvex™ Stealth™ OTG Goggles: Replacement Parts and Accessories
Replacement lenses and accessories for Uvex Stealth OTG goggles
Manufacturer: Honeywell Safety Products S741D
 View more versions of this product
Catalog No. 18-813-430
  Check Availability
Add to Cart
    Specifications
    Type Replacement Lens
    For Use With (Application) Minimizes outdoor sunlight and glare
    Lens Tint Standard Gray
    Surface Coating Dura-streme anti-fog/anti-scratch
    Lens Coating dura-streme anti-fog/anti-scratch
    For use with Uvex Stealth OTG goggles (Cat. No. 18-813-533)
    • Ideal for impact, dust/airborne particles, chemical splash, and optical radiation (UV and welding/heat radiation)
    • Superior optics and panoramic field of vision enhanced with toric lens
    • Pivoting clip and quick-adjusting wide headband
    • Elastomer body conforms and contours for a secure fit—even over most prescription eyewear
    • Replacement lenses easily snap on
  • Description & Specifications
Related Products
Product Content Feedback