Uvex™ Stealth™ OTG Goggles
Low-profile, wraparound style provides complete coverage designed for all-day wear
Manufacturer: Honeywell Safety Products S3971D
 View more versions of this product
Catalog No. 18-813-412
  Check Availability
Add to Cart
    Specifications
    Lens Tint Standard Gray
    Surface Coating Dura-streme anti-fog/anti-scratch
    Lens Coating dura-streme anti-fog/anti-scratch
    Quantity 50/Case
    Type OTG Goggles
    Certifications/Compliance ANSI Z87.1-2010 and CSA Z94.3 standards
    For Use With (Application) Minimizes outdoor sunlight and glare
    Frame Color Navy
    Frame Material Neoprene
  • Description & Specifications
Related Products
Product Content Feedback