Learn More
Honeywell™ Uvex™ Stealth™ Safety Reader Goggles
The Uvex Stealth™ Reader goggle offers an extensive range of magnification strengths. With diopters from +1.0 to +3.0, the Uvex Stealth™ Reader goggle is ideal for workers who require both increased magnification for close-up inspection or fine detail work and the added protection of a goggle. Replacement lenses only.
Manufacturer:  Honeywell™ S751X
 View more versions of this product
Catalog No. 19-066-156
Add to cart
Description
Description
The Uvex Stealth™ continues to be one of the most popular safety goggles on the market and represents the ultimate in modern design, technology and materials. We have again expanded the Uvex Stealth™ family of goggles to include reading magnifiers.
  • Available in Clear with Uvextreme™ anti-fog coating and five diopter strengths from +1.0 to +3.0
  • High quality, soft elastomer body conforms to facial contours for a secure, customized, gap-free fit
  • Goggle body is compatible with Uvex Stealth™ OTG lenses
  • Low profile, wrap-around style provides complete coverage and unobstructed vision complete coverage and protection from chemical splash
  • Wide, adjustable slide headband with pivoting clips provides an easy-to-adjust fit for complete comfort, even with hard hats
  • Economical lens replacement system extends the life of the goggle
  • Meets ANSI Z87.1-2010 (High Impact)
  • Tested by Honeywell to meet the requirements of the CSA Z94.3 standard
  • Made in USA
Specifications
Specifications
2.55mm thick
Uv Extreme Anti-fog/Anti-scratch
+1.50
99.90%
Clear
Blue
Polycarbonate
Safety Reader Goggles
173mm
Safety