Learn More
Uvex™ VersaPro™ Eyewear
Extended-wear comfort features and increased durability
Manufacturer:  Honeywell Safety Products SX0102X
 View more versions of this product
Catalog No. 18-813-448
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Lifetime warranty on frames
Amber
Each
Meets ANSI Z87.1-2003 (High Impact) standard. Certified to the requirements of CSA Z94.3 standard.
Flexible and replaceable nosepiece
Blue; Black
Ratcheting, FlexFit bendable
Wraparound safety glasses
Anti-fog,Scratch-resistant
Ideal for low-light applications where enhanced contrast is needed
Nylon
Polycarbonate
Uvextreme AF
Medium