Uvex™ VersaPro™ Eyewear
Extended-wear comfort features and increased durability
Manufacturer: Honeywell Safety Products SX0152
 View more versions of this product
Catalog No. 18-813-455
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Wraparound safety glasses
    Warranty Lifetime on frames
    Lens Coating Supra-Dura
    Temple Style Ratcheting, FlexFit bendable
    Lens Type Ultra scratch-resistant
    Quantity Each
    For Use With (Application) Ideal for low-light applications where enhanced contrast is needed
    Includes Flexible and replaceable nosepiece
    Frame Color Blue and Black
    Frame Type Large
    Certifications/Compliance Meets ANSI Z87.1-2003 (High Impact) standard. Certified to the requirements of CSA Z94.3 standard.
    Frame Material Nylon
    Lens Material Polycarbonate
    Lens Tint Amber
  • Description & Specifications
Related Products
Product Content Feedback