Learn More
Uvex™ VersaPro™ Eyewear
Extended-wear comfort features and increased durability
Manufacturer:  Honeywell Safety Products SX0100
 View more versions of this product
Catalog No. 18-813-443
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Ultra scratch-resistant; Lifetime warranty on frames
Clear
Each
Meets ANSI Z87.1-2003 (High Impact) standard. Certified to the requirements of CSA Z94.3 standard.
Flexible and replaceable nosepiece
Blue; Black
Ratcheting, FlexFit bendable
Wraparound safety glasses
Scratch-resistant
Ideal for most indoor work applications
Nylon
Polycarbonate
Supra-Dura
Medium