Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Clear UV Transmitting Covers

$179.00 - $248.00

Specifications

Color Clear
Product Type Specs Uv Transmitting Cover
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
UVP19013801
View Documents
UVP
19013801
For use with xx-20 series benchtop uv lamps
Each for $179.00
Add to cart
 
UVP19014001
View Documents
UVP
19014001
For use with xx-15 series benchtop uv lamps
Each for $248.00
Add to cart
 
Description

Description

Specifications

Specifications

Clear
Uv Transmitting Cover
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit