Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UVP Filter Assemblies for White/UV Light Transilluminators
$609.00
Specifications
Product Type | Filter Assembly |
---|---|
Filtration Area (English) | 7.87 x 7.87 in. |
Filtration Area (Metric) | 20 x 20cm |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
UVP38018908
|
UVP
38018908 |
TLW-20 UV/white light transilluminator |
Each for $609.00
|
|
UVP38018905
|
UVP
38018905 |
TMW-20 UV/white light transilluminator |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Specifications
Filter Assembly | |
7.87 x 7.87 in. | |
20 x 20cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title