Learn More
Analytik Jena™ UVP iBox Studio
High quality images and advanced analysis software make this system ideal for labs just starting in vivo applications or in need of a fluorescent in vivo imaging system.
Manufacturer:  Analytik Jena™ 84997093202
 View more versions of this product
Catalog No. UVP97093202
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
  • Powerful and affordable imaging system for high resolution small animal fluorescence imaging
  • Upgradeable for NIR imaging applications or the need of an anesthesia system
  • iBox Studio includes warming plate, keyboard and mouse
Advanced technical specifications and capabilities:
  • Take images of the highest quality with deep cooling, efficient photon-to-signal conversion and high performance cameras. Camera options include either the highest resolution 8.1MP camera or the exceptionally sensitive 3.2MP camera.
  • Wide aperture lens optics, capture more light in low-light applications. Signals come faster and are stronger. This guarantees extremely fast image capture.
  • Multiple illumination sources – RGBW LEDs and NIR lasers make the system extremely versatile
  • Small footprint and integrated 13.3 inch color touchscreen maximize the use of laboratory bench space
  • Included warming plate for up to 3 mice on an ergonomic slide out tray
  • Included GFP and RFP emission filters to enable the most common fluorescence applications
  • Powerful VisionWorks software allows flexibility of creating custom one-touch workflows or using preinstalled one-touch application templates
  • Image acquisition generates unmodified raw data. Users have the choice of applying image enhancement tools.
  • Enhancement and analysis features include many powerful tools such as colony counting, area density, noise reduction, background subtraction, inversion, pseudocolor, compositing and more
  • Applications include in vivo/small animal imaging, fluorescence imaging, NIR imaging, multiplex imaging, plant imaging, and colony counting
  • Optional UVP anesthesia kit available for immobilizing small animals utilizing a combination of oxygen and isofluorane/sevoflurane gas
Specifications
Specifications
8.1MP CCD
50
16 in.
24 in.
18 in.
61 cm
13.3 in.
230 V
5 position filter wheel
8.1 MP
GFP and RFP filter, VisionWorks software
46 cm