Learn More
UVP Sample Plates and Plate Holder for GelMax Imager
Maximize imaging capabilities by adding interchangeable sample plates to view a wide range of fluorophore and colorimetric stains
$259.28 - $731.50
Specifications
For Use With (Application) Maximize imaging capabilities by adding interchangeable sample plates to view a wide range of fluorophore and colorimetric stains
For Use With (Equipment) Gelmax imager
View More Specs
Products 4
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
UVP20165701
View Documents
UVP
20165701
Sample Plate Holder Each for $259.28
Add to cart
UVP38038101
View Documents
UVP
38038101
Longwave Sample Plate Each for $619.00
Add to cart
UVP38038102
View Documents
UVP
38038102
Visi Blue Interchangeable Sample Plate Each for $625.00
Add to cart
UVP38038103
View Documents
UVP
38038103
White Light Sample Plate Each for $731.50
Add to cart
Description
Description
  • For use with the GelMax Imager
  • Plates fit precast and mini gels up to 11.5 x 16cm
Specifications
Specifications
Maximize imaging capabilities by adding interchangeable sample plates to view a wide range of fluorophore and colorimetric stains
Gelmax imager