Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UVRAG Antibody (953035), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9180MFV500
Description
UVRAG Monoclonal antibody specifically detects UVRAG in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| UVRAG | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Cellular Signaling, Epigenetics | |
| 7405 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 953035 | |
| Flow Cytometry, CyTOF-ready | |
| beclin 1 binding protein, DHTX, disrupted in heterotaxy, p63, UV radiation resistance associated gene, UV radiation resistance-associated gene protein, VPS38 | |
| Human UVRAG synthetic peptide corresponding to sequences between amino acids 260-360, Accession # Q9P2Y5 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction