Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UXT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551154
Description
The antibody detects endogenous levels of total UXT protein.
UXT Polyclonal antibody specifically detects UXT in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
UXT | |
Polyclonal | |
Unconjugated | |
UXT | |
0910002B17Rik; androgen receptor trapped clone 27 protein; ART-27; HSPC024; Protein UXT; RP1-212G6.2; SKP2-associated alpha PFD 1; STAP1; ubiquitously expressed prefoldin like chaperone; ubiquitously expressed transcript; ubiquitously expressed transcript protein; ubiquitously-expressed transcript; ubiquitously-expressed, prefoldin-like chaperone; Unknown (protein for MGC:133475); UXT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8409 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UBK9 | |
UXT | |
Full length fusion protein of human UXT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction