Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vβ 10[b] TCR Rat anti-Mouse, BUV395, Clone: B21.5, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 742825

Catalog No. BDB742825


Add to cart

Description

Description

The B21.5 monoclonal antibody specifically recognizes the Vβ10 T-Cell Receptor (TCR) of mice having the b /haplotype (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb /gene complex. It does not react with mice having the a /(eg, C57BR, C57L, SJL, SWR) or c /(eg, RIII) Tcrb /haplotype.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications

Specifications

Vβ 10[b] TCR
B21.5
Brilliant Ultraviolet 395
TCR Vβ10, TCR V beta 10, TCR Vb10
Mouse Cytotoxic T-Cell Clone Soluble αβ TCR
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
Affinity Purified
RUO
Mouse
Rat LOU, also known as Louvain, LOU/C, LOU/M IgG2a λ
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit