Learn More
Vβ 12 T-Cell Receptor Mouse anti-Mouse, BV711, Clone: MR11-1, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 745408
Catalog No. BDB745408
Add to cart
Description
Description
Vβ 12 T-Cell Receptor Monoclonal antibody specifically detects Vβ 12 T-Cell Receptor in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Vβ 12 T-Cell Receptor
MR11-1
Brilliant Violet 711
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Mouse