Learn More
Vγ 2 T-Cell Receptor Hamster anti-Mouse, Brilliant Violet 650, Clone: UC3-10A6, BD Optibuild™
Armenian Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 742311
Catalog No. BDB742311
Add to cart
Description
Description
The UC3-10A6 monoclonal antibody specifically recognizes T-cell Receptor (TCR) V gamma 2 (Vγ2).Vγ2+ TCRγδcells make up significant proportions of TCRγδcells in the late fetal and adult thymus and adult peripheral lymphoid tissues and lung. The frequency of splenicVγ2+ TCRγδcells differs among inbred mouse strains; in C57BL/6 mice, the frequency increases dramatically during the four weeks after birth. Immobilized UC3-10A6 antibody can activateVγ2+TCRγδ cells. Please note that the Vγ2 designation correlates with the nomenclature of Garman, Doherty, and Raulet; the Vγ4 designation of Heiligand Tonegawa is equilvalent.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Specifications
Specifications
Vγ 2 T-Cell Receptor
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
V gamma 2 TCR, Vγ2 TCR, Trgv2
Not Reported
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV650 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
UC3-10A6
Brilliant Violet 650
Affinity Purified
Armenian Hamster
Armenian Hamster IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal