Learn More
Vα2 TCR Rat anti-Mouse, BV421, Clone: B20.1, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566241
Catalog No. BDB566241
Add to cart
Description
Description
Vα2 TCR Monoclonal antibody specifically detects Vα2 TCR in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
Vα2 TCR
B20.1
Brilliant Violet 421
Purified
Rat
Rat LOU, also known as Louvain, LOU/C, LOU/M IgG2a, λ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
T cell receptor alpha, variable region 2
Soluble αβ TCR from mouse cytotoxic T-cell clone KB5-C20
25μg
Cell Biology
Primary
Murine
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.