Learn More
Vβ4 T-Cell Receptor Rat anti-Mouse, Brilliant Violet 711, Clone: KT4, BD Optibuild
Rat Monoclonal Antibody
Manufacturer: BD Biosciences 743023
Description
The KT4 antibody specifically recognizes the Vβ4 T-cell Receptor (TCR) of mice having the a ( e.g /., C57BR, C57L, SJL, SWR), b ( e.g /., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2), and c ( e.g /., RIII) haplotypes of the Tcrb /gene complex. Vβ4 TCR-bearing cells are among the CD4+ autoreactive T cells which induce autoimmune thyroiditis after elimination of regulatory (CD5-bright) T-cell subsets. Plate-bound KT4 antibody activates Vβ4 TCR-bearing T cells.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.Specifications
Vβ4 T-Cell Receptor | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
TCR V beta 4, TCR Vβ4 | |
KT4 B10.D2 mouse T-cell clone I3 | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV711 under optimal conditions that minimize unconjugated dye and antibody. | |
RUO | |
Primary | |
Murine |
KT4 | |
Brilliant Violet 711 | |
Affinity Purified | |
Rat | |
Rat SD, also known as Sprague-Dawley (outbred) IgG2b, κ | |
50μg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
Monoclonal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.