Learn More
Vγ9 TCR Mouse anti-Human, BUV395, Clone: B3, BD Optibuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 744040
Description
The B3 monoclonal antibody specifically recognizes the human variableγ9 subunit (Vγ9) ofγ/δ T- cell receptor (TCR). Theγ/δTCR is composed of two disulfide-linked glycoproteins. The gamma chain is approximately 45-60 kD and is detected on a small subset peripheral blood T cells and thymocytes. The majority of normal peripheral bloodγ/δT cells express a Vγ9+/Vδ2+ phenotype. The reason for this selection in the T-cell repertoire is not well understood.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.Specifications
Vγ9 TCR | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Vgamma9 TCR, V2, TRGC1, TCRGV9, TRGV9 | |
Mouse IgG1, κ | |
50μg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
Monoclonal |
B3 | |
Brilliant Ultraviolet 395 | |
Affinity Purified | |
Murine | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV395 under optimal conditions that minimize unconjugated dye and antibody. | |
RUO | |
Primary | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.