Learn More
TCR V alpha 2 (soluble) Rat anti-Mouse, FITC, Clone: B20.1, Invitrogen™ Non-distribution product as customer accommodation.
Rat Monoclonal Antibody
Manufacturer:  Invitrogen RM5001
Catalog No. 50-113-8179
Add to cart
Description
Description
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
Specifications
Specifications
TCR V alpha 2 (soluble)
Monoclonal
FITC
Liquid
Rat
IgG2a
1 mL
4° C
Primary
Mouse
Flow Cytometry
B20.1
PBS with BSA and 0.1% sodium azide
TCRVA; TCRVA2; V alpha 2; V alpha-2 TCR; Valpha 2; Valpha2; Va 2; Va2
Purified soluble alpha/beta T cell receptor from the cytotoxic Tcell clone, KB5-C20.
Purified
RUO
Antibody
Monoclonal