Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMGL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198264
Description
AMGL Polyclonal specifically detects AMGL in Human samples. It is validated for Western Blot.Specifications
AMGL | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
amelogenin, Y isoform, amelogenin, Y-linked, AMGL, AMGY | |
Rabbit | |
20 kDa | |
100 μL | |
Primary | |
Human: 100%; Dog: 85%; Rat: 85%; Goat: 85%; Horse: 85%; Sheep: 85%; Bovine: 85%; Mouse: 77%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Goat, Sheep | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q99218 | |
AMELY | |
The immunogen for this antibody is AMGL C-terminal region (NP_001134). Peptide Sequence: QPMQPQPPVQPMQPLLPQPPLPPMFPLRPLPPILPDLHLEAWPATDKTKQ | |
Affinity purified | |
RUO | |
266 | |
Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction