Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CP2F2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19826920UL
Description
CP2F2 Polyclonal specifically detects CP2F2 in Rat samples. It is validated for Western Blot.Specifications
CP2F2 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_062176 | |
Rabbit | |
54 kDa | |
20 μL | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Cyp2f2 | |
The immunogen for this antibody is CP2F2 - middle region. Peptide sequence EHQDSLDPNSPRDFIDCFLTKMVQEKQDPLSHFNMDTLLMTTHNLLFGGT. | |
Affinity Purified | |
RUO | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction