Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V alpha 24 J alpha 18 TCR Mouse anti-Human, PE/Atto594, Clone: 6B11, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP200267PEATT594
Description
V alpha 24 J alpha 18 TCR Monoclonal antibody specifically detects V alpha 24 J alpha 18 TCR in Human samples. It is validated for Flow Cytometry.Specifications
V alpha 24 J alpha 18 TCR | |
6B11 | |
Flow Cytometry | |
Conjugated, Purified | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Primary | |
Human |
Flow Cytometry | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
iNKT, iNKT cell, invariant natural killer T cell, invariant NK T cell, invariant NKT cell | |
Peptide epitope in the CDR3 region of V alpha 24 | |
Protein G purified | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title