Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V beta 10b Rat anti-Mouse, PE, Clone: B21.5, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 12580581

 View more versions of this product

Catalog No. 50-108-79


Add to cart

Description

Description

Description: The B21.5 monoclonal antibody reacts with the Vbeta10 T Cell Receptor (TCR) in mice of the b haplotype of the TCR Vbeta gene complex, which includes AKR, BALB/c, CBA, C3H/He, C57BL, DBA/1 and DBA/2 mice. It does not react with Vbeta10 in mice of the a or c haplotypes of the TCR Vbeta gene complex, including C57BR, C57L, SJL, and SWR mice. Applications Reported: This B21.5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B21.5 antibody has been tested by flow cytometric analysis of C57Bl/6 mouse lymph node cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
Specifications

Specifications

TCR V beta 10b
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
B21.5
PE
Tcrb
Tcrb
IgG2a, lambda
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
21577
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.