Learn More
TCR V beta 8.1/8.2 Monoclonal Antibody (KJ16-133 (KJ-16, KJI6, KJ-I6)), FITC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$106.65
Specifications
Antigen | TCR V beta 8.1/8.2 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-987-2
|
Invitrogen
11581382 |
100 μg |
Each of 1
for $106.65
Save 29% Reg : |
|
|||||
Description
Description: This KJ16-133 monoclonal antibody recognizes mouse T cell receptors (TCR) containing Vbeta 8.1 or 8.2, which is expressed by 15-20% of peripheral T cells in most strains of mice (ie. Balb/c, AKR, C57BL/6, B10.A) and by a number of T cell hybridomas (ie. DO-11.10, 3DT-52.5, D1G10G11, SKK-9.11, SKK-45.10, MDK16, S18.4). Applications Reported: This KJ16-133 (KJ-16, KJI6, KJ-I6) antibody has been reported for use in flow cytometric analysis. Applications Tested: This KJ16 (KJ-16, KJI6, KJ-I6) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.Specifications
TCR V beta 8.1/8.2 | |
Flow Cytometry | |
FITC | |
Rat | |
Mouse | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.