Learn More
V Bta 10B TCR Mouse, anti-Mouse, R718, Clone: B21.5, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752205
Catalog No. BDB752205
Add to cart
Description
Description
The B21.5 monoclonal antibody specifically recognizes the Vβ 10 T-Cell Receptor (TCR) of mice having the b haplotype (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb gene complex. It does not react with mice having the a (eg, C57BR, C57L, SJL, SWR) or c (eg, RIII) Tcrb haplotype.
Specifications
Specifications
V Bta 10B TCR
B21.5
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Mouse