Learn More
V Bta 8 TCR Mouse, anti-Human, R718, Clone: JR2, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 751894
Catalog No. BDB751894
Add to cart
Description
Description
The JR2 monoclonal antibody specifically recognizes the β-chain variable region (Vβ8) of the T-cell antigen receptor expressed on 0.5% to 3% of peripheral blood T cells. This antibody has been further characterized as having specificity to a major subset of the BV8 region as determined in the TCR Workshop of July 1995. Staphylococcal enterotoxin E has been shown to specifically stimulate T cells bearing Vβ8 (BV8).
Specifications
Specifications
V Bta 8 TCR
JR2
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human