Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-FIT™ Safety Harness - Back & Shoulder D-Rings, Quick-Connect Leg Straps

The V-FIT™ full body safety harness allows you to focus on your job - not your harness

Supplier:  MSA™ 10195073

 View more versions of this product

Catalog No. 19-001-634


Add to Cart

Description

Description

  • Featuring a patent-pending RaceFLEX™ buckle, bulky chest straps are eliminated for a close and comfortable fit
  • The athletic cut of the V-FIT harness contours to the body, improving upper torso movement on the job, while soft shoulder padding helps to eliminate pressure points and chafing for all day comfort
Specifications

Specifications

Back, Shoulder
25.4 in.
X-Small
64.5 cm
Fall Protection
Vest
181 kg
RaceFLEX™ Quick-Connect Buckle
400 lb.
Quick-Connect
2 lb.
Harness
ANSI Z359.11-2014 CSA Z259.10
Working at Heights
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit