Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-FIT™ Safety Harness - Tongue Buckle Leg Straps, Shoulder & Leg Padding

The V-FIT™ full body safety harness allows you to focus on your job - not your harness

Supplier:  MSA™ 10194977

 View more versions of this product

Catalog No. 19-001-611


Explore available promotions
Add to Cart

Description

Description

  • Featuring a patent-pending RaceFLEX™ buckle, bulky chest straps are eliminated for a close and comfortable fit
  • The athletic cut of the V-FIT harness contours to the body, improving upper torso movement on the job, while soft shoulder padding helps to eliminate pressure points and chafing for all day comfort
Specifications

Specifications

Working at Heights
Back
ANSI Z359.11-2014 CSA Z259.10
Fall Protection
181 kg
25.4 in.
Medium/Large
Tongue
Vest with shoulder and leg padding
400 lb.
RaceFLEX™ Quick-Connect Buckle
64.5 cm
Harness
2 lb.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.