Learn More
V gamma 2 TCR Monoclonal Antibody (UC3-10A6), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Armenian Hamster Monoclonal Antibody
Supplier: Leinco Technologies Inc T353100MG


Description
Specificity: This antibody reacts with Vgamma 2 T-Cell Receptor (TCR)-bearing T lymphocytes. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: This antibody is stable for at least one week when stored sterile at 2-8C. For long term storage aseptically aliquot in working volumes without diluting and store at -20C in a manual defrost freezer. Avoid Repeated Freeze Thaw Cycles.
V gamma 2 TCR Monoclonal antibody specifically detects V gamma 2 TCR in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition AssaysSpecifications
V gamma 2 TCR | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
Armenian Hamster | |
Protein A/G | |
RUO | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays | |
UC3-10A6 | |
Functional Grade | |
TCR V gamma2; TCRV gamma 2; TCRV gamma2; TCRV y2; Vg2; Vgamma2 | |
G8 mouse T cells | |
100 mg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.