Learn More
MSA V-Gard™ Hearing Protection Hygiene Kit
V-Gard™ Hearing Protection Hygiene Kit contains replacement cushions and inserts to maintain the stated sound attenuation performance of your ear muffs. Under normal use, you should replace the hygiene kit at least twice per year.
$21.70
Specifications
Includes 1 Pair of Cushions and 1 Pair of Inserts
Style Hygiene Kit
View More Specs
Products 3
Catalog Number Mfr. No. Noise Reduction Rating (NRR) Price Quantity & Availability  
Catalog Number Mfr. No. Noise Reduction Rating (NRR) Price Quantity & Availability  
19-177-232
View Documents
MSA
10193452
31 dB
Each for $21.70
Add to cart
19-177-233
View Documents
MSA
10193451
27 dB
Each for $21.70
Add to cart
19-177-234
View Documents
MSA
10193450
22 dB
Each for $21.70
Add to cart
Description
Description
  • For proper performance, always use genuine MSA replacement parts
  • Order the proper replacement kit for your specific MSA hearing protector
Specifications
Specifications
1 Pair of Cushions and 1 Pair of Inserts
Hygiene Kit