Learn More
V Gma 2 TCR Mouse, anti-Mouse, R718, Clone: UC3-10A6, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 751923
Catalog No. BDB751923
Add to cart
Description
Description
The UC3-10A6 monoclonal antibody specifically recognizes T-cell Receptor (TCR) V gamma 2 (Vγ2). Vγ2+ TCRγδ cells make up significant proportions of TCRγδ cells in the late fetal and adult thymus and adult peripheral lymphoid tissues and lung. The frequency of splenic Vγ2+ TCRγδ cells differs among inbred mouse strains| in C57BL/6 mice, the frequency increases dramatically during the four weeks after birth. Immobilized UC3-10A6 antibody can activate Vγ2+ TCRγδ cells. Please note that the Vγ2 designation correlates with the nomenclature of Garman, Doherty, and Raulet| the Vγ4 designation of Heiligand Tonegawa is equilvalent.
Specifications
Specifications
V Gma 2 TCR
UC3-10A6
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Mouse