Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ v-Myb Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567636
Description
Predicted to react with Mouse and Rat samples.
A-Myb is expressed at specific times in reproductive tissues, some neural cells, and a subset of normal and neoplastic B lymphocytes. Both A-Myb and B-Myb are expressed in t(14:18) lymphoma cells where they then inhibit cell arrest and apoptotic signaling. Expression of B-Myb rescues cells from p53-induced G1 phase arrest that is mediated by p21, while A-Myb functions as an anti-apoptotic factor by effectively activating the bcl-2 promoter and thereby up-regulating Bcl-2 expression.
Specifications
v-Myb | |
Polyclonal | |
Unconjugated | |
MYBL1 | |
AMYB; A-myb; G1-419-6; MYB proto-oncogene like 1; MYBA; MYBL1; myb-like protein 1; Myb-related protein A; myeloblastosis oncogene-like 1; repro9; VMYB; v-myb avian myeloblastosis viral oncogene homolog-like 1 | |
Rabbit | |
affinity chromatography | |
RUO | |
17864, 297783, 4603 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
P10243, P51960 | |
MYBL1 | |
Synthesized peptide derived from human v-Myb. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction